Order Entry
Austria
ContactUsLinkComponent
N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)-4-nitrobenzamide
N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)-4-nitrobenzamide
Catalog # APOSPC210004-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(2-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)-4-nitrobenzamide
Catalog # APOSPC210004-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :389.72 g/mol
Storage Temperature:Ambient
CAS Number:1610047-56-7
Formula:C₁₅H₁₁ClF₃N₃O₄
MDL Number:MFCD28023539