
Copper(II) hexafluoroacetylacetonate hydrate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:155640-85-0
Formula:C₁₀H₂CuF₁₂O₄·xH₂O
MDL Number:MFCD00151019
:477.64 g/mol (anhy)
Boiling Pt:220 °C
Melting Pt:97…99 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together