
5-Bromo-α,α,α-trifluoro-2-nitrotoluene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:344-38-7
:
Formula:C₇H₃BrF₃NO₂
MDL Number:MFCD00014706
:270.01 g/mol
Boiling Pt:95…100 °C/5mmHg
Melting Pt:33…44 °C
Density:1.799 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together