Potassium-3-fluoro-4-methoxyphenyltrifluoroborate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:850623-62-0
:
Formula:C₇H₆BF₄KO
MDL Number:MFCD04115757
:232.0300 g/mol
Melting Pt:276…280 °C
Storage Temperature:Ambient
Frequently Bought Together