

TBTU (O-(1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium tetrafluoroborate)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:125700-67-6
:
Formula:C₁₁H₁₆BF₄N₅O
MDL Number:MFCD00077413
:321.1 g/mol
Melting Pt:195…205 °C (dec.)
UN:1325
ADR:4.1,II
:GHS02
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together