Order Entry
Austria
ContactUsLinkComponent
(R,R)-(-)-3,6-Octanediol 96%
(R,R)-(-)-3,6-Octanediol 96%
Catalog # APOSOR932401-500MG
Supplier:  Apollo Scientific
CAS Number:  
(R,R)-(-)-3,6-Octanediol 96%
Catalog # APOSOR932401-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :146.23 g/mol
Storage Temperature:Ambient
CAS Number:129619-37-0
Formula:C₈H₁₈O₂
MDL Number:MFCD01631087