trans-1,2-Diaminocyclohexane-N,N,N',N'-tetraacetic acid monohydrate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:125572-95-4
:MFCD00066429
Formula:C₁₄H₂₂N₂O₈·1H₂O
MDL Number:MFCD00149243
:364.35 g/mol
Melting Pt:213…216 °C
Storage Temperature:Ambient
Frequently Bought Together