
(R)-(-)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine 97%
Catalog # APOSOR925455-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:184.24 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:109838-85-9
Formula:C₉H₁₆N₂O₂
MDL Number:MFCD00040565