Order Entry
Austria
ContactUsLinkComponent
Benzaldehyde phenylhydrazone 95%
Benzaldehyde phenylhydrazone 95%
Catalog # APOSOR923751-25G
Supplier:  Apollo Scientific
CAS Number:  
Benzaldehyde phenylhydrazone 95%
Catalog # APOSOR923751-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :196.25 g/mol
Storage Temperature:Ambient
CAS Number:588-64-7
Formula:C₁₃H₁₂N₂
MDL Number:MFCD00051318