Order Entry
Austria
Orders LinkContactUsLinkComponent
(S)-(-)-2-Methoxypropionic acid 97%

(S)-(-)-2-Methoxypropionic acid 97%

Supplier:   Apollo Scientific
(S)-(-)-2-Methoxypropionic acid 97%
Supplier:  Apollo Scientific
Certificates
(S)-(-)-2-Methoxypropionic acid 97%

4 Options Available Below

Product Details & Documents
Synonyms:  (S)-2-Methoxypropanoic acid
  • CAS Number:23953-00-6
  • Formula:C₄H₈O₃
  • MDL Number:MFCD01632589
  • Molecular Weight:104.105 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 33,30 €
    33,30 €
    1 * 1 g
       33,30 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 142,00 €
    142,00 €
    1 * 5 g
       142,00 €

  • Bottle
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g

  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 417,00 €
    417,00 €
    1 * 25 g
       417,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.