Order Entry
Austria
ContactUsLinkComponent
Dimethyl (S)-(-)-methylsuccinate 95%
Dimethyl (S)-(-)-methylsuccinate 95%
Catalog # APOSOR913785-1G
Supplier:  Apollo Scientific
CAS Number:  
Dimethyl (S)-(-)-methylsuccinate 95%
Catalog # APOSOR913785-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :160.17 g/mol
Storage Temperature:Ambient
CAS Number:63163-08-6
Formula:C₇H₁₂O₄
MDL Number:MFCD00134572