Order Entry
Austria
ContactUsLinkComponent
 
(R)-2-Isopropylamino-2-phenylethanol 95%
Catalog # APOSOR912554-1G
Supplier:  Apollo Scientific
CAS Number:  
(R)-2-Isopropylamino-2-phenylethanol 95%
Catalog # APOSOR912554-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :179.26 g/mol
Storage Temperature:Ambient
CAS Number:112211-92-4
Formula:C₁₁H₁₇NO
MDL Number:MFCD00671548