5-(Hydroxymethyl)-2-furaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:67-47-0
:
Formula:C₆H₆O₃
MDL Number:MFCD00003234
Boiling Pt:114…116 °C/1mmHg
Melting Pt:32…35 °C
Density:1.243 g/cm³ (25 °C)
Flash Pt:79 °C
:126.11 g/mol
EINECS:200-654-9
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together