8-Hydroxyquinoline-N-Oxide
Catalog # APOSOR8255-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
Melting Pt:138…140 °C
:161.16 g/mol
Storage Temperature:Ambient
CAS Number:1127-45-3
Formula:C₉H₇NO₂
MDL Number:MFCD00006739