Order Entry
Austria
ContactUsLinkComponent
2-[4-(4-Nitrophenyl)-1,3-thiazol-2-yl]acetonitrile 97%
2-[4-(4-Nitrophenyl)-1,3-thiazol-2-yl]acetonitrile 97%
Catalog # APOSOR8141-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[4-(4-Nitrophenyl)-1,3-thiazol-2-yl]acetonitrile 97%
Catalog # APOSOR8141-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :245.26 g/mol
Melting Pt:147…148 °C
Storage Temperature:Ambient
CAS Number:69625-13-4
Formula:C₁₁H₇N₃O₂S
MDL Number:MFCD00724226