
Cesium chloride 99.99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7647-17-8
:
Formula:CsCl
MDL Number:MFCD00010955
:168.36 g/mol
Melting Pt:646 °C
Density:3.988 g/cm³ (25 °C)
EINECS:231-600-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together