
2-(4-(tert-Butyl)phenyl)acetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3288-99-1
:
Formula:C₁₂H₁₅N
MDL Number:MFCD00128112
Boiling Pt:141…142 °C/13mmHg
Density:0.962 g/cm³ (25 °C)
UN:3276
ADR:6.1,III
:173.2500 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together