Geranyl acetate 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (E)-3,7-Dimethylocta-2,6-dien-1-yl acetate
CAS Number:105-87-3
:
Formula:C₁₂H₂₀O₂
MDL Number:MFCD00015037
Boiling Pt:138 °C/25mmHg
Density:0.916 g/cm³ (25 °C)
Molecular Weight:196.29 g/mol
Flash Pt:104 °C
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together