Potassium xanthogenate (potassium-O-ethyl dithiocarbonate)
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:140-89-6
:
Formula:C₃H₅KOS₂
MDL Number:MFCD00004931
:160.30 g/mol
Melting Pt:210 °C
Density:1.558 g/cm³ (25 °C)
Flash Pt:96 °C
Storage Temperature:Ambient
UN:3342
ADR:4.2,III
:GHS02
:GHS07
Frequently Bought Together