
2,3-Dichlorophenylhydrazinium chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:21938-47-6
:
Formula:C₆H₇Cl₃N₂
MDL Number:MFCD00052265
Melting Pt:∼ 204 °C (dec.)
UN:2811
ADR:6.1,III
:213.49 g/mol
:GHS06
Storage Temperature:Refrigerator
Frequently Bought Together