(R)-(-)-2-Methylpiperazine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:75336-86-6
:
Formula:C₅H₁₂N₂
MDL Number:MFCD00192317
:100.16 g/mol
Boiling Pt:155…156 °C
Melting Pt:86…93 °C
Flash Pt:65 °C
Storage Temperature:Ambient
UN:2925
ADR:4.1+8,III
:GHS02
:GHS05
Frequently Bought Together