
2-(Aminosulphonyl)-N,N-dimethylnicotinamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:112006-75-4
:
Formula:C₈H₁₁N₃O₃S
MDL Number:MFCD03265507
:229.26 g/mol
EINECS:413-440-7
:GHS07
Storage Temperature:Ambient
Frequently Bought Together