5-Chloro-3-methylbenzo[b]thiophene-2-sulphonyl chloride
Catalog # APOSOR5377-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:281.18 g/mol
Melting Pt:153 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
CAS Number:166964-33-6
Formula:C₉H₆Cl₂O₂S₂
MDL Number:MFCD00173797