
(1R,4R)-2-Oxa-5-azabicyclo[2.2.1]heptane hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:601515-79-1
:
Formula:C₅H₁₀ClNO
MDL Number:MFCD01321291
:135.59 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together