
Ethyl-4-hydroxyquinoline-2-carboxylate 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:24782-43-2
:
Formula:C₁₂H₁₁NO₃
MDL Number:MFCD01863732
:217.2200 g/mol
Melting Pt:216…218 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together