
2-(Methylsulphonyl)aniline hydrochloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:205985-95-1
:
Formula:C₇H₁₀ClNO₂S
MDL Number:MFCD00216484
Melting Pt:139…141 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together