
(R)-1-Methyl-3-pyrrolidinol 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:104641-60-3
:
Formula:C₅H₁₁NO
MDL Number:MFCD03788747
:101.15 g/mol
Boiling Pt:103…106 °C/35mmHg
Density:0.921 g/cm³ (25 °C)
Flash Pt:70 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together