
2-Bromo-4-isopropylaniline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:51605-97-1
:
Formula:C₉H₁₂BrN
MDL Number:MFCD00051585
:214.1 g/mol
Boiling Pt:116…118 °C/3mmHg
Density:1.35 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS07
Frequently Bought Together