Order Entry
Austria
ContactUsLinkComponent
 
Irritant
(R)-(-)-3-Aminopiperidine dihydrochloride
Catalog # APOSOR4587-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(R)-(-)-3-Aminopiperidine dihydrochloride
Catalog # APOSOR4587-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :173.08 g/mol
Melting Pt:190…195 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:334618-23-4
Formula:C₅H₁₄Cl₂N₂
MDL Number:MFCD06799458