Order Entry
Austria
ContactUsLinkComponent
2-Methylpropene
2-Methylpropene
Catalog # APOSOR40492-100G
Supplier:  Apollo Scientific
CAS Number:  
2-Methylpropene
Catalog # APOSOR40492-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Cylinder
  • Pk:
    100 g
  • Seq:
    3401
  • Environmentally Preferable:
  • Pk Info:
    1/4" NPTM connection

Specifications

About this item

Synonyms: 2-Methylpropene, Isobutene, Isobutylene
 :
 :56.11
Melting Pt:–140 °C
EINECS:204-066-3
UN:1055
 :GHS02
 :GHS04
Storage Temperature:Ambient
CAS Number:115-11-7
Formula:C₄H₈
MDL Number:MFCD00008898
Boiling Pt:–6.9 °C
Density:0.5879 g/cm³ (25 °C)
Flash Pt:–80 °C
ADR:2.1
Merck Index:12,05155