
1-Acetylnaphthalene 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:941-98-0
:
Formula:C₁₂H₁₀O
MDL Number:MFCD00004013
:170.21 g/mol
Boiling Pt:301…303 °C
Melting Pt:10.5 °C
Density:1.12 g/cm³ (25 °C)
Flash Pt:>110 °C
EINECS:213-384-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together