2-(2-Formyl-4-methoxyphenoxy)acetonitrile 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:149384-09-8
:
Formula:C₁₀H₉NO₃
MDL Number:MFCD02082011
:191.19 g/mol
Storage Temperature:Raumtemperatur
Frequently Bought Together