Order Entry
Austria
ContactUsLinkComponent
2-(9,10-Dioxo-9,10-dihydro-2-anthracenyl)acetonitrile 95%
2-(9,10-Dioxo-9,10-dihydro-2-anthracenyl)acetonitrile 95%
Catalog # APOSOR32993-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(9,10-Dioxo-9,10-dihydro-2-anthracenyl)acetonitrile 95%
Catalog # APOSOR32993-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.26 g/mol
Storage Temperature:Ambient
CAS Number:121831-04-7
Formula:C₁₆H₉NO₂
MDL Number:MFCD01315214