Dimethyl phenylphosphonate
Catalog # APOSOR322065-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:186.15 g/mol
Storage Temperature:Ambient
CAS Number:2240-41-7
Formula:C₈H₁₁O₃P
MDL Number:MFCD00059292