
Ethyl-4-hydroxyquinoline-3-carboxylate 98%
Catalog # APOSOR307983-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:217.2206 g/mol
Melting Pt:271 °C
:GHS07
Storage Temperature:Ambient
CAS Number:26892-90-0
Formula:C₁₂H₁₁NO₃
MDL Number:MFCD00173406