(6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinolin-1-yl)acetonitrile
Catalog # APOSOR307544-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.28 g/mol
Storage Temperature:Ambient
CAS Number:52244-06-1
Formula:C₁₃H₁₆N₂O₂
MDL Number:MFCD00800509