
(1R,2R)-(-)-1,2-Diaminocyclohexane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:20439-47-8
:
Formula:C₆H₁₄N₂
MDL Number:MFCD00062985
:114.19 g/mol
Boiling Pt:104…110 °C/40mmHg
Melting Pt:40…43 °C
Flash Pt:70 °C
Storage Temperature:Ambient
UN:3259
ADR:8,II
:GHS05
Frequently Bought Together