
5-Bromo-2-methoxynicotinic acid 97%
Catalog # APOSOR30675-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:232.04 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:54916-66-4
Formula:C₇H₆BrNO₃
MDL Number:MFCD06656771