Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4-nitrobenzamide
2-Chloro-4-nitrobenzamide
2-Chloro-4-nitrobenzamide
APOSOR30292-1G
:
Apollo Scientific
:
2-Chloro-4-nitrobenzamide
APOSOR30292-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
200.58 g/mol
:
GHS07
:
Ambient
:
3011-89-0
:
C₇H₅ClN₂O₃
:
MFCD00017119