2-Bromo-1-(3-methylbenzo[b]thiophen-2-yl)ethan-1-one
Catalog # APOSOR26021-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:269.17 g/mol
:GHS05
Storage Temperature:Ambient
CAS Number:118337-33-0
Formula:C₁₁H₉BrOS
MDL Number:MFCD00173793