
1-[2-(2,5-Dimethylpyrrol-1-yl)ethyl]piperazine
Catalog # APOSOR25971-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:207.32 g/mol
Boiling Pt:178…180 °C/0.2mmHg
Melting Pt:12 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
CAS Number:5059-36-9
Formula:C₁₂H₂₁N₃
MDL Number:MFCD03407319