(1-Methyl-1H-pyrazol-3-yl)methanol 97%
Catalog # APOSOR2057-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: 3-(Hydroxymethyl)-1-methyl-1H-pyrazole
:
:112.13 g/mol
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS07
CAS Number:84547-62-6
Formula:C₅H₈N₂O
MDL Number:MFCD08690274