Order Entry
Austria
ContactUsLinkComponent
3-Bromopyridine-N-oxide 97+%
Corrosive
Irritant
3-Bromopyridine-N-oxide 97+%
Catalog # APOSOR2026-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
3-Bromopyridine-N-oxide 97+%
Catalog # APOSOR2026-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :173.9954 g/mol
Boiling Pt:130 °C/2mmHg
Melting Pt:30…40 °C
Flash Pt:>110 °C
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:2402-97-3
Formula:C₅H₄BrNO
MDL Number:MFCD01646056