Order Entry
Austria
ContactUsLinkComponent
4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoic acid
Irritant
4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoic acid
Catalog # APOSOR16087-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Methyl-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzoic acid
Catalog # APOSOR16087-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :306.3187 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:641569-94-0
Formula:C₁₇H₁₄N₄O₂
MDL Number:MFCD11521324