Order Entry
Austria
ContactUsLinkComponent
6-(tert-Butyl)-3-methyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
Irritant
6-(tert-Butyl)-3-methyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
Catalog # APOSOR15389-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-(tert-Butyl)-3-methyl-2-amino-4,7-dihydrothieno[2,3-c]pyridine-3,6(5H)-dicarboxylate
Catalog # APOSOR15389-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :312.39 g/mol
Melting Pt:165…166 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:877041-47-9
Formula:C₁₄H₂₀N₂O₄S
MDL Number:MFCD06200952