Caffeic acid
New Product
Anbieter: Apollo Scientific
Jump to:
About this item
Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
- CAS Number:331-39-5
- Formula:C₉H₈O₄
- MDL Number:MFCD00004392
- Molecular Weight:180.16 g/mol
- Melting Pt:194…198 °C
- Storage Temperature:Ambient
- EINECS:206-361-2
6 Options Available Below
Product Details & Documents
Synonyms: 3,4-Dihydroxycinnamic acid, 3-(3,4-Dihydroxyphenyl)acrylic acid
- CAS Number:331-39-5
- Formula:C₉H₈O₄
- MDL Number:MFCD00004392
- Molecular Weight:180.16 g/mol
- Melting Pt:194…198 °C
- Storage Temperature:Ambient
- EINECS:206-361-2
Product Family Options
Product Information
- Pack typeVerfügbarkeitPreis
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



