Potassium acetate 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:127-08-2
:
Formula:CH₃COOK
MDL Number:MFCD00012458
:98.14 g/mol
Melting Pt:303…307 °C
Density:1.57 g/cm³ (25 °C)
EINECS:204-822-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together