Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Acetyl-2-naphthol
1-Acetyl-2-naphthol 97%
1-Acetyl-2-naphthol 97%
APOSOR015967-25G
:
Apollo Scientific
:
1-Acetyl-2-naphthol 97%
APOSOR015967-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
186.21 g/mol
:
130…132 °C/2mmHg
:
63…67 °C
:
209-369-4
:
GHS07
:
Ambient
:
574-19-6
:
C₁₂H₁₀O₂
:
MFCD00012131