2-Adamantanone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:700-58-3
:
Formula:C₁₀H₁₄O
MDL Number:MFCD00074737
:150.22 g/mol
Melting Pt:257 °C (subl.)
EINECS:211-847-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Toluene, EMPLURA®, extra pure, Supelco®
Supplier: Merck
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥99.8% stabilised, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
N,N-Dimethylformamide 99.0-100.00%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
n-Heptane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
2-Propanol ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
1,6-Diaminohexane ≥98%
Supplier: Thermo Fisher Scientific