Order Entry
Austria
Orders LinkContactUsLinkComponent
Copper(I) thiocyanate

Copper(I) thiocyanate

Irritant
Environment
Supplier:   Apollo Scientific
Copper(I) thiocyanate
Supplier:  Apollo Scientific
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Copper thiocyanate, Copper (I) rhodanide, Cuprous thiocyanate, Copper rhodanide
  • CAS Number:1111-67-7
  • Formula:CCuNS
  • MDL Number:MFCD00010980
  • Molecular Weight:121.63 g/mol
  • Melting Pt:1084 °C
  • Density:2.84 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:214-183-1
  • UN:3077
  • ADR:9+EHS,III
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 46,40 €
    46,40 €
    1 * 50 g
       46,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSIN9847-50G
  • Bottle
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 171,00 €
    171,00 €
    1 * 250 g
       171,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    APOSIN9847-250G
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.